missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) bromide hydrate

Product Code. p-7048875
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11381858

Brand: Thermo Scientific Alfa Aesar 011816.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123333-87-9 | Br3Rh | 342.62 g/mol

Rhodium(III) bromide hydrate is used to prepare coordination complex, trans-dibromotetrakis(pyridine)rhodium(III) bromide hexahydrate. It is also involved in the preparation of rhodium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-87-9
Molecular Formula Br3Rh
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149837
InChI Key MMRXYMKDBFSWJR-UHFFFAOYSA-K
IUPAC Name rhodium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Rh+3]

Specifications

Physical Form Crystalline
Quantity 1 g
Formula Weight 342.62 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Rhodium(III) bromide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.