missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) bromide hydrate

Product Code. 11391858
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391858 5 g 5g
11381858 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391858 Supplier Thermo Scientific Alfa Aesar Supplier No. 011816.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 123333-87-9 | Br3Rh | 342.62 g/mol

Rhodium(III) bromide hydrate is used to prepare coordination complex, trans-dibromotetrakis(pyridine)rhodium(III) bromide hexahydrate. It is also involved in the preparation of rhodium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 123333-87-9
Molecular Formula Br3Rh
Molecular Weight (g/mol) 342.62
MDL Number MFCD00149837
InChI Key MMRXYMKDBFSWJR-UHFFFAOYSA-K
IUPAC Name rhodium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Rh+3]

Spécification

Physical Form Crystalline
Quantity 5 g
Formula Weight 342.62 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Rhodium(III) bromide hydrate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.