missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min, Thermo Scientific Chemicals

Catalog Number 11390709
Click to view available options
Quantity:
0.5 g
1 g
5 g
14284-92-5
C15H21O6Rh
400.23
MFCD00083144 MFCD00083144
DGOINFUDFBWCMX-LNTINUHCSA-K
rhodium iii acetylacetonate-rh
131675882
[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
14284-92-5
C15H21O6Rh
400.23
MFCD00083144 MFCD00083144
DGOINFUDFBWCMX-LNTINUHCSA-K
rhodium iii acetylacetonate-rh
131675882
[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Rhodium(III) 2,4-pentanedionate is used in catalytic reagents for organic synthesis. Rhodium(III) 2,4-pentanedionate is useful for hydrosilylation of a variety of organic substrates like alkenes, terminal or internal acetylenes, conjugated dienes, alpha, beta -unsaturated aldehydes and allylic compounds. It performs the role of a catalyst in well-known reactions such as transformation processes including hydrogenation, ethanol steam reforming, CO oxidation, and NOx reduction. Rhodium acetylacetonate, as the organic solvent-soluble source of rhodium metal ions, can be used to prepare monodisperse size/shape controlled Rh nanocrystals. Rh nanocrystal monolayers have been reported to show high activity for hydrogenation of vinylcyclohexene to selectively produce ethylcyclohexene. The size/shape controlled nanocrystals find a variety of applications in oil refining processes, chemical synthesis and fuel cell technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 263°C to 264°C
Physical Form Crystalline
Quantity 1 g
Solubility Information Very soluble in chloroform; Soluble in alcohol; In soluble in water.
IUPAC Name (E)-4-hydroxypent-3-en-2-one;rhodium
Formula Weight 400.23
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Rh 25.2% min.
Assay (metals basis)
Chemical Name or Material Rhodium(III) 2,4-pentanedionate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'