missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh

Product Code. 10402781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402781 1 g Glass bottle 1g
10062580 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402781 Supplier Thermo Scientific Acros Supplier No. 197610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29998-99-0 | C8H16O10Rh2 | 478.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29998-99-0
Molecular Formula C8H16O10Rh2
Molecular Weight (g/mol) 478.02
MDL Number MFCD00149212
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium

Specifications

Color Green
Physical Form Fine Powder
Quantity 1 g
Linear Formula [(CH3CO2)2RhH2O]2
Fieser 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in many polar solvents like eth, anol, acetone, acetic acid, acetonitrile, dmso, ni, tromethane, thf, triethyl phosphate.
Formula Weight 478.02
Percent Purity ∼43% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.