missing translation for 'onlineSavingsMsg'
Learn More

Rhodium on activated carbon, 5% Rh, unreduced, dry

Product Code. 10378140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10378140 5 g Glass bottle 5g
10754733 1 g Glass bottle 1g
10511261 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10378140 Supplier Thermo Scientific Acros Supplier No. 195440050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-16-6 | Rh | 102.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6 , 7440-44-0
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Black to Black-Gray
Quantity 5 g
Packaging Glass bottle
Particle Size Typical particle size distribution: D50: 20 Micron, <8 Micron: 17%, <16 Micron: 39%,< 32 Micron: 67%, <64 Micron: 90%, <128 Micron: 99%
Form Powder
Fieser 01,982; 03,326; 04,418; 06,503; 08,433
Merck Index 15, 8310
Formula Weight 102.91
Percent Purity 5% (Rh)
Chemical Name or Material Rhodium on activated carbon, 5% Rh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.