missing translation for 'onlineSavingsMsg'
Learn More

Rhodium acetate, ca. 40% Rh, brown, water soluble, Thermo Scientific Chemicals

Product Code. 10125070
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42204-14-8
Molecular Formula C2H3O2Rh
Molecular Weight (g/mol) 161.95
MDL Number MFCD03410291
InChI Key NENDHUHGFRLXEN-UHFFFAOYSA-M
Synonym rhodium 3+ ion acetate
PubChem CID 3660445
IUPAC Name acetic acid;rhodium
SMILES [Rh].CC([O-])=O

Specifications

Color Brown
Physical Form Powder
Quantity 1 g
Linear Formula Rh(CO2CH3)x
Solubility Information Solubility in water: soluble
Formula Weight 130.96
Percent Purity ∼40% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium acetate, ca. 40% Rh

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'