missing translation for 'onlineSavingsMsg'
Learn More

Rhodium acetate, ca. 40% Rh, brown, water soluble

Product Code. 10247413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10247413 5 g 5g
10125070 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10247413 Supplier Thermo Scientific Acros Supplier No. 270200050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 42204-14-8 | C2H3O2Rh | 161.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42204-14-8
Molecular Formula C2H3O2Rh
Molecular Weight (g/mol) 161.95
MDL Number MFCD03410291
InChI Key NENDHUHGFRLXEN-UHFFFAOYSA-M
Synonym rhodium 3+ ion acetate
PubChem CID 3660445
IUPAC Name acetic acid;rhodium
SMILES [Rh].CC([O-])=O

Specifications

Color Brown
Physical Form Powder
Quantity 5 g
Linear Formula Rh(CO2CH3)x
Solubility Information Solubility in water: soluble
Formula Weight 130.96
Percent Purity ∼40% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium acetate, ca. 40% Rh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.