missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Rhodium, 5% on activated carbon paste, C101023-5

Brand:  Alfa Aesar™ 044863.04

Code : NEW

Additional Details : CAS Number : 7440-16-6 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11326279

  • £46.60 / 2g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Rhodium
7440-16-6
5% on activated carbon paste, C101023-5
MFCD03458392

Specifications

Rhodium
7440-16-6
MFCD03458392
2g
5% on activated carbon paste, C101023-5
Paste
Black
Videos
SDS