missing translation for 'onlineSavingsMsg'
Learn More

Rhodium 10%, Palladium 1% on activated carbon powder, Thermo Scientific Chemicals

Product Code. 11480670
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11480670

Brand: Thermo Scientific Alfa Aesar 044124.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN1325
Odor Odorless
Chemical Name or Material Rhodium, 10%, Palladium 1% on activated carbon powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.