Learn More
Rhodamine B base, 97%, Thermo Scientific Chemicals
Rhodamine B base, CAS # 509-34-2, is a fluorescent dye used in staining applications.
£51.80 - £133.00
Chemical Identifiers
CAS | 509-34-2 |
---|---|
Molecular Formula | C28H30N2O3 |
MDL Number | MFCD00066967 |
Synonym | C.I. 45170.1, Solvent Red 49 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10378250
|
Thermo Scientific Acros
215230250 |
25 g | Glass bottle |
£51.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10701165
|
Thermo Scientific
215231000 |
100 g | Glass bottle |
£133.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
509-34-2 | |
C28H30N2O3 | |
MFCD00066967 | |
C.I. 45170.1, Solvent Red 49 |
Specifications
509-34-2 | |
C28H30N2O3 | |
19, 344 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dimethylformamide,, dimethylsulfoxide, ethanol | |
442.55 | |
97% | |
Fine Crystalline Powder |
99% | |
MFCD00066967 | |
C.I. 45170.1, Solvent Red 49 | |
Authentic | |
1% max. (105°C) | |
Lilac | |
Rhodamine B base |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Suspected of causing genetic defects.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Rhodamine B base
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only