missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B, 98+%

Product Code. 13428498
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10, C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point 210.0°C to 211.0°C
Assay Percent Range ≥98%
Beilstein 19, V,8, 669
Merck Index 15, 8307
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol
Infrared Spectrum Authentic
Formula Weight 479.01
Percent Purity ≥98%
Grade Pure
Packaging Plastic Drum
Vapor Pressure Vapor Pressure: Negligible
Decomposition Information >255°C
Color Green
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Rhodamine B, Pure
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Rhodamine B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'