missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B

Product Code. 11460084
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460084 1000 g 1000g
11470084 50 g 50g
11480084 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460084 Supplier Thermo Scientific Chemicals Supplier No. A13572.0B
  • ONLINE EXCLUSIVE PRICE
    £265.20
    £216.65 / 1000g
    Save £48.55
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suitable for fluorescent labellingRhodamine B is used as a laser dye, metal chelating agent and an additive to drugs and cosmetics. It also serves as a staining fluorescent dye, which is used as a colorant in inks, paper and plastics. Further, it is used to make pink fluorescent pigments. It is also employed as a biomarker in oral rabies vaccines for wildlife such as raccoons. In addition to this, it is a useful fluorochrome for histology, fluorescence resonance energy transfer (FRET) and mitochondrial probe.

Solubility
Soluble in water, benzene and ethanol. Slightly soluble in acetone, hydrochloric acid, sodium hydroxide, methyl cellosolve and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10; C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Melting Point ∼210°C (decomposition)
Beilstein 4119648
Density 1.31
Merck Index 14,8183
Solubility Information Soluble in water,benzene and ethanol. Slightly soluble in acetone,hydrochloric acid,sodium hydroxide,methyl cellosolve and alcohol.
Quantity 1000 g
Formula Weight 479.02
Chemical Name or Material Rhodamine B
compliance-icons compliance-icons
Product Identifier
  • Rhodamine B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Food Red 15

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.