missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine B, pure

Product Code. p-2692129 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10396870

Brand: Thermo Scientific Chemicals 132311000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-88-9
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.02
MDL Number MFCD00011931
InChI Key PYWVYCXTNDRMGF-UHFFFAOYSA-N
Synonym Basic Violet 10, C.I. 45170
PubChem CID 6694
ChEBI CHEBI:52334
IUPAC Name [9-(2-carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium;chloride
SMILES [Cl-].CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=CC=C3C(O)=O)=C2C=C1)N(CC)CC

Specifications

Absorbance 1700 (at lambda min.)
Melting Point 210.0°C to 211.0°C
Absorption Ratio 1.1 to 1.6
Beilstein 19, V,8, 669
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in hcl and naoh, soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 549 to 558nm (in 50:50 IMS:water)
Formula Weight 479.01
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Rhodamine B
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Rhodamine B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.