missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G, pure

Product Code. 10286580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10286580 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10286580 Supplier Thermo Scientific Chemicals Supplier No. 191410500
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 290.0°C
CAS 989-38-8
Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies
Molecular Formula C28H31ClN2O3
MDL Number MFCD00012665
Beilstein 18, V,12, 283
Synonym Basic Red 1, C.I. 45160
Color Brown to Brown/Red
Solubility Information Solubility in water: soluble.
Quantity 50 g
InChI Key VYXSBFYARXAAKO-UHFFFAOYSA-N
Physical Form Fine Crystalline Powder
SMILES [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
Molecular Weight (g/mol) 479.017
PubChem CID 51358423
Formula Weight 479.01
Grade Pure
Chemical Name or Material Rhodamine 6G
Show More Show Less
compliance-icons
Product Identifier
  • Rhodamine 6G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.