missing translation for 'onlineSavingsMsg'
Learn More

Rhodamine 110, TRC

Product Code. 30424824
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Product Code. Quantity
30424824 100 mg
30413235 250 mg
30427571 1 g
3 options
This item is not returnable. View return policy

Product Code. 30424824

Brand: Toronto Research Chemicals TRCR318580100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13558-31-1
Formula Weight 366.0771
InChI Formula InChI=1S/C20H14N2O3.ClH/c21-11-5-7-15-17(9-11)25-18-10-12(22)6-8-16(18)19(15)13-3-1-2-4-14(13)20(23)24;/h1-10,21H,22H2,(H,23,24);1H
IUPAC Name 2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride
Molecular Formula C20 H14 N2 O3 . Cl H
Molecular Weight (g/mol) 366.8
SMILES Cl.Nc1ccc2C(=C3C=CC(=N)C=C3Oc2c1)c4ccccc4C(=O)O
Synonym 3,6-Diamino-9-(2-carboxyphenyl)xanthylium Chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic Acid Monohydrochloride, 3,6-Diamino-9-(2-carboxyphenyl)xanthylium Chloride, R 110, RH 110, Rhodamine 560, Rhodamine N
Chemical Name or Material Rhodamine 110

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.