missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RhoD Antibody (1E7), Novus Biologicals™

Product Code. 18344629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344629 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344629 Supplier Novus Biologicals™ Supplier No. H00029984M12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RhoD Monoclonal antibody specifically detects RhoD in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RhoD
Applications ELISA
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01338
Gene Alias ARHDRHOM, ras homolog D, ras homolog gene family, member A, ras homolog gene family, member D, Rho, RhoD, RhoHP1, rho-related GTP-binding protein RhoD, Rho-related protein HP1
Host Species Mouse
Immunogen RHOD (AAH01338, 1 a.a. ∽ 210 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTAAQAAGEEAPPGVRSVKVVLVGDGGCGKTSLLMVFADGAFPESYTPTVFERYMVNLQVKGKPVHLHIWDTAGQDDYDRLRPLFYPDASVLLLCFDVTSPNSFDNIFNRWYPEVNHFCKKVPIIVVGCKTDLRKDKSLVNKLRRNGLEPVTYHRGQEMARSVGAVAYLECSARLHDNVHAVFQEAAEVALSSRGRNFWRRITQGFCVVT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29984
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.