missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rhod-3 Calcium Imaging Kit

Product Code. 10013833
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10013833 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10013833 Supplier Thermo Scientific™ Supplier No. R10145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Rhod-3 Calcium Imaging Kit

The novel Rhod-3 Calcium Imaging Kit is designed for live-cell imaging of cytosolic calcium signaling in combination with green-fluorescent dyes or proteins.The red-fluorescent Rhod-3 AM dye supplied in the Rhod-3 Calcium Imaging Kit offers significant improvements over existing Ca2+-sensing dyes. The uptake into organelles has been reduced, which translates into better cytosolic localization. Also, Rhod-3 AM exhibits a large increase in fluorescence emission upon binding to calcium, making it more sensitive than other red-fluorescent calcium dyes.

  • Compared to existing red calcium dyes, Rhod-3 AM exhibits a large increase (>2.5 fold) in fluorescence upon binding Ca2+ and very low fluorescence at rest (without Ca2+ binding).
  • Narrow emission enables simultaneous detection with GFP or other dyes.
  • Enhanced cytosolic localization for better detection of cytosolic transients

For easy cell loading, the Rhod-3 Calcium Imaging Kit contains PowerLoad™ Concentrate. Due to the unique nature of the PowerLoad™ solution, it can be used in the presence of complete culture media, thus reducing the negative effects of replacing media or loading in serum-free media.

A proprietary, water-soluble form of probenecid (which is commonly used to inhibit cellular transport and thus reduces the baseline signal) is also supplied with the Rhod-3 Calcium Imaging Kit.

Calcium indicators are important tools in signal transduction and cell-based pharmacological screening. In cells and tissues with blue or green autofluorescence, long-wavelength (i.e., red-shifted) calcium indicators provide a means to bypass this overlapping autofluorescence. Additionally, red-shifted calcium dyes are well suited for calcium imaging experiments multiplexed with green-fluorescent protein (GFP) or other green fluorescent dyes.

The Rhod-3 AM dye (included in the Rhod-3 Calcium Imaging Kit) is an improved red-shifted rhodamine-based calcium dye with an attached AM ester group. Once inside the cell, intracellular esterases cleave the AM group, trapping the dye in the cell and rendering the dye fluorescent. As calcium binds, the fluorescence intensity of the dye is enhanced. The spectral properties of Rhod-3 AM (ex/em = ∼560/600 nm) and strong fluorescence upon calcium binding provide an excellent signal window and make it our brightest red calcium dye. Additionally, Rhod-3 AM displays a more uniform cytosolic distribution and improved signal compared to existing red calcium dyes such as Rhod-2.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage The kit contains sufficient reagents for staining 50 coverslips based upon the protocol provided. Store kit at -20°C, desiccated and protected from light. When stored as directed, this kit is stable for 1 year.
Detection Method Fluorescence
For Use With (Application) Calcium Assay
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Dye Type Fluorescent Dye-Based
Emission Visible
Form Lyophilized
Modification Un-Modified, Un-Modified
Product Line Molecular Probes
Quantity 1 Kit
Shipping Condition Room Temperature
Sub Cellular Localization Cytoplasm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.