missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoC Antibody (OTI3A5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73888H
This item is not returnable.
View return policy
Description
RhoC Monoclonal specifically detects RhoC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RhoC | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RHOC | |
| Synthetic peptide: VFEMATRAGLQVRKNKRRRG, corresponding to C terminal of human RhoC. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI3A5 | |
| Western Blot, Immunohistochemistry | |
| ARH9ARHCRho cDNA clone 9, H9, MGC1448, MGC61427, oncogene RHO H9, ras homolog gene family, member C, RAS-related homolog 9, RhoC, rhoC GTPase, RHOH9, rho-related GTP-binding protein RhoC, small GTP binding protein RhoC | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 389.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction