missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RhoBTB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116836
This item is not returnable.
View return policy
Description
Positive Samples: Rat lung Immunogen sequence: MDLSEGELGG PSEPGGTHPE DHQGHSDQHH HHHHHHHGRD FLLRAASFDV CESVDEAGGS GPAGLRASTS DGILRGNGTG YLPGRGRVLS SWSRAFVSIQ EEMAEDPLTY KSRLMVVVKM DSSIQPGPFR AVLKYLYTGE LDENERDLMH IAHIAELLEV F.
RHOBTB2 is a member of the evolutionarily conserved RHOBTB subfamily of Rho GTPases. For background information on RHOBTBs, see RHOBTB1.
Specifications
| RhoBTB2 | |
| Polyclonal | |
| Unconjugated | |
| RHOBTB2 | |
| Dbc2; Deleted in breast cancer 2 gene protein; deleted in breast cancer 2 gene protein homolog; E130206H14Rik; KIAA0717; KIAA0740; mKIAA0717; p83; Rho related BTB domain containing 2; Rho related BTB domain containing protein 2; RHOBTB1; RHOBTB2; Rho-related BTB domain containing 2; rho-related BTB domain-containing protein 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23221, 246710, 306004 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.46 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91V93, Q9BYZ6 | |
| RHOBTB2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-480 of human RHOBTB2 (NP_0011535081). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction