missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | RHOBTB2 |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | DBC2/3361 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | Q9BYZ6 |
| Gene Alias | DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?