missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RhoBTB2 Monoclonal Antibody (2G6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521429
This item is not returnable.
View return policy
Description
Peptide Sequence: TISAHKPLLI SSCDWMAAMF GGPFVESSTR EVVFPYTSKS CMRAVLEYLY TGMFTSSPDL DDMKLIILAN RLCLPHLVAL TEQYTVTGLM EATQMMVDID GDVLVFLELA.
RHOBTB2 is a member of the evolutionarily conserved RHOBTB subfamily of Rho GTPases. For background information on RHOBTBs, see RHOBTB1.
Specifications
| RhoBTB2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BYZ6 | |
| RHOBTB2 | |
| RHOBTB2 (AAH34917, 510 a.a. ~ 619 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 2G6 | |
| Unconjugated | |
| RHOBTB2 | |
| Dbc2; Deleted in breast cancer 2 gene protein; deleted in breast cancer 2 gene protein homolog; E130206H14Rik; KIAA0717; KIAA0740; mKIAA0717; p83; Rho related BTB domain containing 2; Rho related BTB domain containing protein 2; RHOBTB1; RHOBTB2; Rho-related BTB domain containing 2; rho-related BTB domain-containing protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23221 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction