missing translation for 'onlineSavingsMsg'
Learn More

RHOBTB2 Antibody (DBC2/4570), Novus Biologicals™

Mouse Monoclonal Antibody

£178.00 - £351.00

Specifications

Antigen RHOBTB2
Clone DBC2/4570
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664085
View Documents
Novus Biologicals
NBP3-23791-100ug
100 μg
£351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18628844
View Documents
Novus Biologicals
NBP3-23791-20ug
20 μg
£178.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RHOBTB2 Monoclonal antibody specifically detects RHOBTB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

RHOBTB2
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Signal Transduction
10 mM PBS with 0.05% BSA
23221
IgG2 κ
Protein A or G purified
DBC2/4570
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
DBC2KIAA0717Deleted in breast cancer 2 gene protein, p83, Rho-related BTB domain containing 2, rho-related BTB domain-containing protein 2
Human recombinant RHOBTB2 protein fragment (around aa 554-604) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.