missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoB Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94104-0.1ml
This item is not returnable.
View return policy
Description
RhoB Polyclonal antibody specifically detects RhoB in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RhoB | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| ARH6MSTP081, ARHBAplysia RAS-related homolog 6, h6, MST081, oncogene RHO H6, ras homolog gene family, member B, Rho cDNA clone 6, RHOH6RhoB, rho-related GTP-binding protein RhoB | |
| A synthetic peptide corresponding to a sequence within amino acids 100-190 of human RhoB (NP_004031.1). VPEVKHFCPNVPIILVANKKDLRSDEHVRTELARMKQEPVRTDDGRAMAVRIQAYDYLECSAKTKEGVREVFETATRAALQKRYGSQNGCI | |
| 0.1 mL | |
| Angiogenesis | |
| 388 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction