missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RHOA/CEN3q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | ras homolog gene family, member A |
Gene Alias | ARH12,ARHA,RHO12,RHOH12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073355
|
Abnova™
FG0220.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083355
|
Abnova™
FG0220.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
ARH12,ARHA,RHO12,RHOH12 | |
387 | |
Liquid |
Tube | |
ras homolog gene family, member A | |
RHOA | |
DNA/RNA |