missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RhoA Monoclonal antibody specifically detects RhoA in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | RhoA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1A11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH01360 |
| Gene Alias | h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?