missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoA Antibody (101), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90106AF350
This item is not returnable.
View return policy
Description
RhoA Monoclonal specifically detects RhoA in Human samples. It is validated for ELISA.
Specifications
| RhoA | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 101 | |
| ELISA | |
| h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human RhoA (P61586-1; Met1-Leu193). | |
| 0.1 mL | |
| Cancer, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 387 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction