missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rho Pan Polyclonal Antibody, Invitrogen™
Description
Glycerol (1:1) may be added for additional stability.
Specifications
Specifications
| Antigen | Rho Pan |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | whole serum with no preservative |
| Gene | RHOQ |
| Gene Accession No. | P08134, P17081, P61586, P62745, Q9HBH0, P62746, Q62159, Q8BYP3, Q8R527, Q9QUI0, P61589, P62747, Q9JJL4 |
| Gene Alias | Rho-related GTP-binding protein RhOA, RhOB, RhOC, RhOF, RhOQ, RAC1, RAC2, RAC3, CDC42 |
| Gene Symbols | RHOA, RHOB, RHOC, RHOF, RHOQ |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?