missing translation for 'onlineSavingsMsg'
Learn More

Rho Pan Polyclonal Antibody, Invitrogen™

Product Code. 11511262
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11511262 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11511262 Supplier Invitrogen Supplier No. OSR00297G

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glycerol (1:1) may be added for additional stability. Reconstitute in 500 μL of sterile water

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rho Pan
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with no preservative
Gene RHOQ
Gene Accession No. P08134, P17081, P61586, P62745, Q9HBH0, P62746, Q62159, Q8BYP3, Q8R527, Q9QUI0, P61589, P62747, Q9JJL4
Gene Alias Rho-related GTP-binding protein RhOA, RhOB, RhOC, RhOF, RhOQ, RAC1, RAC2, RAC3, CDC42
Gene Symbols RHOA, RHOB, RHOC, RHOF, RHOQ
Host Species Rabbit
Immunogen A synthetic peptide from human/rat/mouse/chicken/dog/frog
Purification Method Ammonium sulfate precipitation
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104215, 117273, 11848, 11852, 11853, 23433, 23912, 295342, 387, 388, 389, 54509, 64373, 690130, 85428
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.