missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rho A/B/C Antibody (SR1355), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27831-50ul
This item is not returnable.
View return policy
Description
Rho A/B/C Monoclonal antibody specifically detects Rho A/B/C in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Rho A/B/C | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 387 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR1355 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Aplysia ras-related homolog 12, Aplysia RAS-related homolog 6, ARH12, ARH6, ARH9, ARHA, ARHB, ARHC, EDFAOB, epididymis secretory sperm binding protein, h6, H9, MST081, MSTP081, oncogene RHO H12, oncogene RHO H6, oncogene RHO H9, ras homolog gene family, member B, ras homolog gene family, member C, RAS-related homolog 9, Rho cDNA clone 6, RHO12, rhoC GTPase, RHOH12, RHOH6, RHOH9, rho-related GTP-binding protein RhoB, rho-related GTP-binding protein RhoC, small GTP binding protein RhoA, small GTP binding protein RhoC, transforming protein RhoA | |
| A synthesized peptide derived from human Rho A/B/C (Uniprot #: P61586/P62745/P08134 ) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction