missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rheumatoid Factor Monoclonal Antibody (RF2-17-109.2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110592
This item is not returnable.
View return policy
Description
This antibody reacts with 30% of IgM-RF from unrelated individuals. It does not recognize IgM, IgA, or IgG proteins lacking RF nor with F(ab’)2 fragments.
Antibodies directed against the Fc fragment of immunoglobulin G (IgG) are called rheumatoid factors (RFs). They are heterogenous and are usually IgM antibodies. RFs are used as a marker in individuals with suspected rheumatoid arthritis (RA) or other autoimmune conditions. The normal reference range for RF is less than 15 IU/mL.
Specifications
| Rheumatoid Factor | |
| Monoclonal | |
| 1.8 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA | |
| RF2-17-109.2 | |
| Unconjugated | |
| RF | |
| Purified proteins from patients with IgM cryoglobulins. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido