missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhesus Macaque TNF-alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £474.00
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | 182312 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733019
![]() |
R&D Systems
MAB10702-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159260
![]() |
R&D Systems
MAB10702 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rhesus Macaque samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| TNF-alpha | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Rhesus Macaque | |
| P48094 | |
| 7124 | |
| E. coli-derived recombinant rhesus macaque TNF-alpha Val77-Leu233 Accession # P48094 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rhesus macaque TNF-alpha in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse (rm) EDA, rmFas Ligand, rmOX40, rcrTNF-alpha , rhTNF-alpha , rmTNF-alpha , rpTNF-alpha , rrTNF-alpha , rmTRAIL, or rmTWEAK. |
| 182312 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| TNF | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title