Learn More
Thermo Scientific™ Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Brand:  Thermo Scientific™ 7025
Code : 16
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10776524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Specifications
Specifications
Sample Loop
Rheodyne Injector Models 7010/7125
0.76 mm
For Rheodyne 7010/7125; 200 uL; 0.76 mm ID
0.030 in.
200 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.