Learn More
Thermo Scientific™ Rheodyne™ Sample Loops
Shop All Thermo Scientific ProductsBeschreibung
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Spezifikation
Spezifikation
| Type | Sample Loop |
| For Use With (Equipment) | Rheodyne Injectors 7010/7125 |
| Diameter (English) Inner | 0.020 in. |
| Diameter (Metric) Inner | 0.51 mm |
| Volume (Metric) | 50 μL |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.