missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(VII) oxide, 99.9% (metals basis)

Product Code. 11308438
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308438 2 g 2g
11398428 10 g 10g
11388428 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11308438 Supplier Thermo Scientific Alfa Aesar Supplier No. 040468.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a catalyst for oxidation/reduction reactions | CAS: 1314-68-7 | O7Re2 | 484.407 g/mol

As a catalyst for oxidation/reduction reactionsRhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-68-7
Molecular Formula O7Re2
Molecular Weight (g/mol) 484.407
MDL Number MFCD00011199
InChI Key NBGOSNNAAHDRLK-UHFFFAOYSA-N
Synonym rhenium heptoxide, rhenium vii oxide, re2o7, dirhenium heptoxide, dirhenium heptaoxide, rhenium oxide re2o7, trioxo-trioxorheniooxy rhenium, trioxo trioxorheniooxy rhenium, rhenium vii oxide-re, rhenium vii oxide trace metals basis
PubChem CID 123106
IUPAC Name trioxo(trioxorheniooxy)rhenium
SMILES O=[Re](=O)(=O)O[Re](=O)(=O)=O

Specifications

Boiling Point 360°C
Melting Point 297°C
Color Green to Yellow
Physical Form Crystalline
Quantity 2 g
Linear Formula Re2O7
UN Number UN3085
Merck Index 14,8178
Solubility Information Dissolves in water to form HReO4; Soluble in alkalis and acids; Soluble in water and alcohol.
Formula Weight 484.4
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 6.103 g/mL
Assay (metals basis)
Chemical Name or Material Rhenium(VII) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.