missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(V) chloride, 99.9% (metals basis)

Product Code. p-7058809
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11329428

Brand: Thermo Scientific Alfa Aesar 043294.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13596-35-5 | Cl5Re | 363.457 g/mol

A starting material for rhenium porphyrins and alkyne complexes of rhenium. It is a laboratory reagent that can be used as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13596-35-5
Molecular Formula Cl5Re
Molecular Weight (g/mol) 363.457
MDL Number MFCD00011197
InChI Key XZQYTGKSBZGQMO-UHFFFAOYSA-I
Synonym rhenium pentachloride, rhenium v chloride, rhenium chloride recl5, recl5, rhenium 5+ pentachloride, acmc-209c4t
PubChem CID 83602
IUPAC Name pentachlororhenium
SMILES Cl[Re](Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 2 g
Solubility Information Reacts with water.
Formula Weight 363.47
Percent Purity 99.9%
Odor Acidic
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Rhenium (V) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.