missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium(V) chloride, 99.9% (metals basis)
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Description
A starting material for rhenium porphyrins and alkyne complexes of rhenium. It is a laboratory reagent that can be used as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | (decomposition) |
| Physical Form | Powder |
| Quantity | 10 g |
| Solubility Information | Reacts with water. |
| Formula Weight | 363.47 |
| Percent Purity | 99.9% |
| Odor | Acidic |
| Sensitivity | Moisture sensitive |
| Assay | (metals basis) |
| Chemical Name or Material | Rhenium (V) chloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction