missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium Standard: Re @ 10000 μg/mL in 5% HNO3
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Specifications
Specifications
| Chemical Name or Material | Rhenium |
| CAS | 7440-15-5 |
| Analyte or Component Names | Rhenium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Re |
| Concentration | 10000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Synonym | Re |
| InChI Formula | InChI=1 S/Re |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction