missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium Standard for ICP, ICP-MS 10,000 µg/mL (10,000 ppm) in Water (H2O), Reagecon
Manufactured from ultra-pure metal (99.999%) tested by an ISO17025 Accredited Test Method (INAB Ref: 264T) and raw materials assayed by titration and ICP-MS prior to manufacture.
Brand: Reagecon PRE4B7
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- ICP, ICP-MS Reference Standard, ICP, ICP-MS Reference Materia
- ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
- ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
- ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
- ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
- ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
- GHS/OSHA compliant, REACH etc.
- This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
- The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
- All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications
Matrix: H2O | |
HDPE Triple-leached | |
Rhenium | |
Liquid |
10,000 µg/mL (10,000 ppm) | |
250 mL | |
ICP, ICP-MS, AA, IC, XRF and other cation measurement techniques |