missing translation for 'onlineSavingsMsg'
Learn More

Rhenium powder, 99.99%, Thermo Scientific Chemicals

Catalog Number 11331418
Click to view available options
Quantity:
0.2 g
2 g
10 g
50 g
This item is not returnable. View return policy
7440-15-5
Re
186.21
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
CHEBI:49882
rhenium
[Re]
This item is not returnable. View return policy
7440-15-5
Re
186.21
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
CHEBI:49882
rhenium
[Re]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Appearance Powder
Assay Percent Range 99.99% (Metals basis)
Quantity 50 g
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
Mesh Size -325 Mesh
Form Powder
Chemical Name or Material Rhenium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'