missing translation for 'onlineSavingsMsg'
Learn More

Rhenium(IV) sulfide, 99%

Product Code. 11433790
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433790 2 g 2g
11423790 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433790 Supplier Thermo Scientific Alfa Aesar Supplier No. 089482.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12038-63-0 | ReS2 | 250.33 g/mol

Rhenium(IV) sulfide is used as a diamagnetic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12038-63-0
Molecular Formula ReS2
Molecular Weight (g/mol) 250.33
MDL Number MFCD00016307
InChI Key USWJSZNKYVUTIE-UHFFFAOYSA-N
Synonym rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
PubChem CID 82864
IUPAC Name bis(sulfanylidene)rhenium
SMILES S=[Re]=S

Specifications

Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water. Soluble in 1N sodium hydroxide solution.
Formula Weight 250.33
Percent Purity 99%
Odor Odorless
Density 7.506 g/mL
Chemical Name or Material Rhenium (IV) sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.