missing translation for 'onlineSavingsMsg'
Learn More

Rhein, TRC

Product Code. 30424438
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424438 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424438 Supplier Toronto Research Chemicals Supplier No. TRCR31850050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rhein
CAS 478-43-3
Purity Grade Notes HPLC
Molecular Formula C15 H8 O6
Quantity 50 mg
Synonym 2-Anthracenecarboxylic acid','9,10-dihydro-4,5-dihydroxy-9,10-dioxo-, 2-Anthraquinonecarboxylic acid','4,5-dihydroxy- (6CI), 2-Anthroic acid','9,10-dihydro-4,5-dihydroxy-9,10-dioxo- (8CI), 1,8-Dihydroxy-3-carboxyanthraquinone, 1,8-Dihydroxyanthraquinone-3-carboxylic acid, 4,5-Dihydroxy-2-anthraquinonecarboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid, Cassic acid, Chrysazin-3-carboxylic acid, Monorhein, NSC 38629, Rheic acid, Rhein, Rheinic acid, Rhubarb yellow
InChI Formula InChI=1S/C15H8O6/c16-9-3-1-2-7-11(9)14(19)12-8(13(7)18)4-6(15(20)21)5-10(12)17/h1-5,16-17H,(H,20,21)
SMILES OC(=O)c1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1
IUPAC Name 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
Molecular Weight (g/mol) 284.22
Formula Weight 284.0321
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.