missing translation for 'onlineSavingsMsg'
Learn More

Rhein, MedChemExpress

Product Code. 16462346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462346 500 mg 500mg
16441486 200 mg 200mg
16418986 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16462346 Supplier Medchem Express Supplier No. HYN0105
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rhein is a lipophilic anthraquinone extensively found in medicinal herbs, and has many pharmacological effects, including epatoprotective, nephroprotective, anti-inflammatory, antioxidant, anticancer, and antimicrobial activities.

Rhein is a lipophilic anthraquinone extensively found in medicinal herbs, and has many pharmacological effects, including epatoprotective, nephroprotective, anti-inflammatory, antioxidant, anticancer, and antimicrobial activities.

Specifications

Chemical Name or Material Rhein
CAS 478-43-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H8O6
Quantity 500 mg
Synonym Rheic Acid Rhubarb yellow Monorhein
Solubility Information DMSO : 12.17 mg/mL (42.82 mM; Need ultrasonic and warming)
SMILES O=C(C(C=C1C2=O)=CC(O)=C1C(C3=C2C=CC=C3O)=O)O
Molecular Weight (g/mol) 284.22
Formula Weight 284.22
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.