missing translation for 'onlineSavingsMsg'
Learn More

Rhein, 97%

Product Code. 15487408
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487408 250 mg 250mg
15477408 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487408 Supplier Thermo Scientific Alfa Aesar Supplier No. H60902.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 478-43-3 | C15H8O6 | 284.223 g/mol

Rhein is used to inhibit growth factor beta-1 induced plasminogen activator inhibitor-1 in endothelial cells. It acts as an antibacterial agent against Staphylococcus aureus. It is also used as a laxative. Further, it is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhein is used to inhibit growth factor beta-1 induced plasminogen activator inhibitor-1 in endothelial cells. It acts as an antibacterial agent against Staphylococcus aureus. It is also used as a laxative. Further, it is employed as a pharmaceutical intermediate.

Solubility
Soluble in sodium hydroxide. Insoluble in water, dimethylsulfoxide and ethanol.

Notes
Incompatible with oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 478-43-3
Molecular Formula C15H8O6
Molecular Weight (g/mol) 284.223
MDL Number MFCD00009618
InChI Key FCDLCPWAQCPTKC-UHFFFAOYSA-N
Synonym rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid
PubChem CID 10168
ChEBI CHEBI:8825
IUPAC Name 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 2222155
Solubility Information Soluble in sodium hydroxide. Insoluble in water,dimethylsulfoxide and ethanol.
Formula Weight 284.22
Percent Purity 98%
Chemical Name or Material Rhein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.