missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ Rhein

Product Code. 18768886
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18768886 10 mg 10mg
18725986 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18768886 Supplier Tocris™ Supplier No. 6867/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FTO (mRNA N6-methyladenine demethylase) inhibitor; also inhibits ALKBH2 and ALKBH3

Rhein is a FTO (mRNA N6-methyladenine demethylase) inhibitor (IC50 = 1.2 - 21 μM; Ki = 1.8 μM). Competitively binds enzyme at active site preventing binding of N6-methyladenosine (m6A) substrate and resulting in accumulation of DNA damage in cells. Also inhibits ALKBH3 and ALKBH2 (IC50 values are 5.3 and 9.1 μM, respectively). Sensitizes glioblastoma cells to alkylating agents. Active in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C15H8O6
Formula Weight 284.22
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.