missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHEB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520129
This item is not returnable.
View return policy
Description
A suggested positive control is mouse brain tissue lysate. PA5-20129 can be used with blocking peptide PEP-0247.
Rheb encodes a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. This protein is vital in regulation of growth and cell cycle progression due to its role in the insulin/TOR/S6K signaling pathway. The protein has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form, and farnesylation of the protein is required for this activity. Three pseudogenes have been mapped, two on chromosome 10 and one on chromosome 22.
Specifications
| RHEB | |
| Polyclonal | |
| Unconjugated | |
| RHEB | |
| GTP-binding protein Rheb; Ras homolog enriched in brain; Ras homolog enriched in brain 2; RAS-homolog enriched in brain; RHEB; Rheb1; RHEB2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19744, 6009 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q15382, Q921J2 | |
| RHEB | |
| A 15 amino acid peptide from the middle region of human Rheb. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction