missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rheb Antibody (Rheb 5D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50273-0.025ml
This item is not returnable.
View return policy
Description
Rheb Monoclonal antibody specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Rheb | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunoprecipitation 5 μg/mL | |
| GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
| Human Rheb GST fusion | |
| 0.025 mL | |
| Cancer, mTOR Pathway, Signal Transduction | |
| 6009 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| Rheb 5D5 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction