missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rheb Antibody (Rheb 5D5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50273B
This item is not returnable.
View return policy
Description
Rheb Monoclonal specifically detects Rheb in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Rheb | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| RHEB | |
| Human Rheb GST fusion | |
| 0.1 mL | |
| Cancer, mTOR Pathway, Signal Transduction | |
| 6009 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot | |
| Rheb 5D5 | |
| Western Blot | |
| GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction