missing translation for 'onlineSavingsMsg'
Learn More

Rheb Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18406274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406274 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406274 Supplier Novus Biologicals Supplier No. NBP177214AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rheb Polyclonal antibody specifically detects Rheb in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rheb
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from the middle region of human Rheb. The immunogen is located within amino acids 40 - 90 of Rheb. Amino Acid Squence: DTAGQDEYSIFPQTY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6009
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.