missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rheb Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76432AF532
This item is not returnable.
View return policy
Description
Rheb Polyclonal antibody specifically detects Rheb in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Rheb | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 532 | |
| GTP-binding protein Rheb, MGC111559, Ras homolog enriched in brainRHEB2Ras homolog enriched in brain 2 | |
| Antibody was raised against a 14 amino acid synthetic peptide from the amino terminus of human Rheb. The immunogen is located within the first 50 amino acids of Rheb. Amino Acid Squence: PQSKSRKIAILGYR | |
| 0.1 mL | |
| Cancer, mTOR Pathway, Signal Transduction | |
| 6009 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction