missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RHD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107014
This item is not returnable.
View return policy
Description
RHD Polyclonal Antibody for Western Blot
The Rhesus (Rh) blood group system represents one of the most complex and important systems in humans. Two highly homologous genes RHD and RHCE (collectively referred to as RH30 or RHCED) encode the antigens of the Rh blood group system (1-4). These tightly linked genes map to human chromosomal position 1p34.1-1p36. The RHD gene, which is commonly deleted from a large segment of the population, encodes the most potent blood group immunogen, the D antigen (2,5). Rh incompatibility between maternal and fetal blood types results in hemolytic disease of the newborn (HDN), which often results in fetal death (1,2,6). The RHCE gene exists in four allelic forms, and each allele determines the expression of two antigens in Ce, ce, cE, or CE combinations (2,4,5). The RHCED antigens exist as integral membrane proteins with contain 12-transmembrane helices, and maintain erythrocyte membrane integrity. The presentation of the Rh antigenic activity requires the formation of a complex between the RHCED antigens and RHAG (RH50).
Specifications
| RHD | |
| Polyclonal | |
| Unconjugated | |
| RHD | |
| blood group antigen D; blood group protein RHD; blood group Rh(D) polypeptide; CD240D; D antigen (DCS); DIIIc; Erythrocyte membrane glycoprotein Rh30; Rh; Rh blood group antigen Evans; Rh blood group D antigen; Rh blood group protein; Rh blood group, D anitgen; Rh blood group, D antigen; RH polypeptide 2; Rh30; Rh4; Rhced; RHD; RhD blood group antigen; RhD polypeptide; RhDCw; RHDel; RHDVA(TT); Rhesus blood group CE and D; Rhesus blood group D antigen allele DIII type 7; rhesus D antigen; Rhesus system D polypeptide; RhII; RhK562-II; Rhl1; Rh-like polypeptide; RhPI; RHPII; RHXIII; truncated rhesus blood group D antigen; truncated rhesus D | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q02161 | |
| RHD | |
| Synthetic peptide located within the following region: WALTLEAALI LLFYFFTHYD ASLEDQKGLV ASYQVGQDLT VMAAIGLGFL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction